Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108855.1 | NP_001102325.1 | 5 - 6 | 1721 | 84 | |
XM_008769486.1 | XP_008767708.1 | 4 - 5 | 844 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |