Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106513.1 | NP_001099983.1 | 4 - 5 | 1063 | 116 | |
XM_008762162.1 | XP_008760384.1 | 4 - 5 | 1008 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |