Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762061.1 | XP_008760283.1 | 7 - 8 | 1209 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |