Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011955.1 | NP_001011955.1 | 1 - 2 | 128 | 95 | |
GenBank mRNA | BC086551.1 | - | 1 - 2 | 128 | 7302418 | 95 |
FQ211655.1 | - | 1 - 2 | 457 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |