Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011955.1 | NP_001011955.1 | 2 - 3 | 258 | 68 | |
XM_006234549.2 | XP_006234611.1 | 1 - 2 | 1099 | 68 | ||
XM_008762023.1 | XP_008760245.1 | 2 - 3 | 207 | 68 | ||
GenBank mRNA | BC086551.1 | - | 2 - 3 | 258 | 7302418 | 68 |
FQ211655.1 | - | 2 - 3 | 587 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |