Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106511.1 | NP_001099981.1 | 7 - 8 | 963 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |