Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109594.1 | NP_001103064.1 | 5 - 6 | 814 | 77 | |
XM_008762035.1 | XP_008760257.1 | 4 - 5 | 842 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |