Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108588.1 | NP_001102058.1 | 9 - 10 | 1538 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |