Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc20a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031148.1 | NP_112410.1 | 6 - 7 | 1110 | 84 | |
XM_006234991.2 | XP_006235053.1 | 6 - 7 | 1334 | 84 | ||
GenBank mRNA | AB000489.1 | - | 6 - 7 | 1150 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |