Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012717.1 | NP_036849.1 | 7 - 8 | 707 | 135 | |
XM_006234438.2 | XP_006234500.1 | 7 - 8 | 962 | 135 | ||
XM_006234439.2 | XP_006234501.1 | 6 - 7 | 800 | 135 | ||
GenBank mRNA | L27487.1 | - | 7 - 8 | 980 | 135 | |
X70658.1 | - | 7 - 8 | 707 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |