Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serping1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199093.1 | NP_954524.1 | 4 - 5 | 957 | 118 | |
XM_006234447.2 | XP_006234509.1 | 5 - 6 | 1032 | 118 | ||
GenBank mRNA | BC061860.1 | - | 4 - 5 | 957 | 5598525 | 118 |
FQ219227.1 | - | 3 - 4 | 661 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |