Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133405.2 | NP_596896.2 | 8 - 9 | 1349 | 151 | |
GenBank mRNA | AY033877.1 | - | 8 - 9 | 1359 | 151 | |
BC166416.1 | - | 8 - 9 | 1349 | 9033484 | 151 | |
FQ211347.1 | - | 8 - 9 | 1359 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |