Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata5l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109647.1 | NP_001103117.1 | 2 - 3 | 1316 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |