Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014258.1 | NP_001014280.1 | 2 - 3 | 574 | 110 | |
XM_006235078.2 | XP_006235140.1 | 2 - 3 | 768 | 110 | ||
XM_006235079.2 | XP_006235141.1 | 2 - 3 | 757 | 110 | ||
XM_006235080.2 | XP_006235142.1 | 2 - 3 | 257 | 110 | ||
GenBank mRNA | BC085849.1 | - | 2 - 3 | 574 | 7189638 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |