Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108584.1 | NP_001102054.1 | 1 - 2 | 359 | 59 | |
XM_006234528.2 | XP_006234590.1 | 1 - 2 | 750 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |