Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047089.1 | NP_001040554.1 | 3 - 4 | 375 | 76 | |
XM_008762089.1 | XP_008760311.1 | 2 - 3 | 2286 | 76 | ||
GenBank mRNA | AB121675.1 | - | 3 - 4 | 375 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |