Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037358.2 | NP_001032435.2 | 2 - 3 | 451 | 63 | |
NM_001244779.1 | NP_001231708.1 | 2 - 3 | 341 | 63 | ||
NM_001244780.1 | NP_001231709.1 | 2 - 3 | 316 | 63 | ||
NM_001244781.1 | NP_001231710.1 | 2 - 3 | 346 | 63 | ||
GenBank mRNA | BC105772.1 | - | 2 - 3 | 451 | 7930561 | 63 |
CK476274.1 | - | 2 - 3 | 302 | 63 | ||
CO559731.1 | - | 316 | 63 | |||
DY319476.1 | - | 2 - 3 | 342 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |