Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053963.2 | NP_446415.2 | 1 - 2 | 130 | 79 | |
GenBank mRNA | BC088120.1 | - | 1 - 2 | 130 | 7380302 | 79 |
X98517.1 | - | 1 - 2 | 130 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |