Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013095.1 | NP_001013113.1 | 10 - 11 | 951 | 73 | |
GenBank mRNA | BC086383.1 | - | 10 - 11 | 951 | 7301572 | 73 |
FQ213005.1 | - | 10 - 11 | 992 | 73 | ||
FQ225883.1 | - | 10 - 11 | 984 | 73 | ||
FQ226276.1 | - | 10 - 11 | 983 | 73 | ||
FQ229703.1 | - | 10 - 11 | 982 | 73 | ||
FQ230888.1 | - | 10 - 11 | 977 | 73 | ||
FQ232595.1 | - | 10 - 11 | 981 | 73 | ||
FQ232661.1 | - | 10 - 11 | 981 | 73 | ||
FQ234018.1 | - | 10 - 11 | 985 | 73 | ||
FQ234727.1 | - | 8 - 9 | 833 | 73 | ||
FQ234921.1 | - | 10 - 11 | 982 | 73 | ||
FQ234929.1 | - | 10 - 11 | 984 | 73 | ||
FQ235264.1 | - | 7 - 8 | 765 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |