Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013095.1 | NP_001013113.1 | 2 - 3 | 72 | 55 | |
GenBank mRNA | BC086383.1 | - | 2 - 3 | 72 | 7301572 | 55 |
FQ213005.1 | - | 2 - 3 | 113 | 55 | ||
FQ225883.1 | - | 2 - 3 | 105 | 55 | ||
FQ226276.1 | - | 2 - 3 | 104 | 55 | ||
FQ229703.1 | - | 2 - 3 | 103 | 55 | ||
FQ230888.1 | - | 2 - 3 | 98 | 55 | ||
FQ232595.1 | - | 2 - 3 | 102 | 55 | ||
FQ234018.1 | - | 2 - 3 | 105 | 55 | ||
FQ234921.1 | - | 2 - 3 | 103 | 55 | ||
FQ234929.1 | - | 2 - 3 | 105 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |