Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106800.1 | NP_001100270.1 | 3 - 4 | 541 | 105 | |
XM_006242487.2 | XP_006242549.1 | 4 - 5 | 810 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |