Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108124.3 | NP_001101594.2 | 4 - 5 | 656 | 72 | |
XM_006242523.2 | XP_006242585.1 | 4 - 5 | 770 | 72 | ||
XM_006242524.2 | XP_006242586.1 | 4 - 5 | 952 | 72 | ||
GenBank mRNA | BC079415.1 | - | 4 - 5 | 628 | 72 | |
BC168198.1 | - | 4 - 5 | 647 | 7113061 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |