Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108127.1 | NP_001101597.1 | 1 - 2 | 71 | 109 | |
XM_006242558.2 | XP_006242620.1 | 1 - 2 | 694 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |