Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106799.1 | NP_001100269.1 | 1 - 2 | 143 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |