Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106799.1 | NP_001100269.1 | 6 - 7 | 941 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |