Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728514.4 | XP_002728560.1 | 11 - 12 | 1903 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |