Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107368.2 | NP_001100838.1 | 4 - 5 | 358 | 127 | |
XM_008771871.1 | XP_008770093.1 | 4 - 5 | 3243 | 127 | ||
GenBank mRNA | BC168209.1 | - | 4 - 5 | 358 | 7133190 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |