Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013058.1 | NP_001013076.1 | 2 - 3 | 256 | 60 | |
XM_006254205.2 | XP_006254267.1 | 2 - 3 | 363 | 60 | ||
GenBank mRNA | BC079248.1 | - | 2 - 3 | 256 | 7132972 | 60 |
FQ230702.1 | - | 1 - 2 | 115 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |