Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033698.1 | NP_001028870.1 | 2 - 3 | 617 | 91 | |
GenBank mRNA | BC099837.1 | - | 2 - 3 | 617 | 7936269 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |