Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253100.2 | XP_006253162.1 | 5 - 6 | 2165 | 64 | |
XM_008771246.1 | XP_008769468.1 | 5 - 6 | 1168 | 64 | ||
XM_008771247.1 | XP_008769469.1 | 5 - 6 | 763 | 64 | ||
XM_008771248.1 | XP_008769470.1 | 5 - 6 | 2008 | 64 | ||
GenBank mRNA | FQ215658.1 | - | 5 - 6 | 607 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |