Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253139.2 | XP_006253201.1 | 1 - 2 | 370 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |