Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047086.1 | NP_001040551.1 | 4 - 5 | 478 | 87 | |
XM_006253139.2 | XP_006253201.1 | 4 - 5 | 739 | 87 | ||
XM_008771256.1 | XP_008769478.1 | 4 - 5 | 574 | 87 | ||
GenBank mRNA | BC166593.1 | - | 4 - 5 | 519 | 7934031 | 87 |
DQ674266.1 | - | 4 - 5 | 478 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |