Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017443.1 | NP_001017443.1 | 6 - 7 | 759 | 108 | |
XM_006253141.2 | XP_006253203.1 | 6 - 7 | 675 | 108 | ||
XM_006253143.2 | XP_006253205.1 | 7 - 8 | 1756 | 108 | ||
XM_008771257.1 | XP_008769479.1 | 5 - 6 | 561 | 108 | ||
GenBank mRNA | BC092575.1 | - | 6 - 7 | 759 | 7114719 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |