Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191073.1 | NP_001178002.1 | 1 - 2 | 140 | 87 | |
GenBank mRNA | AY724480.1 | - | 259 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |