Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771266.1 | XP_008769488.1 | 5 - 6 | 1064 | 138 | |
XM_008771267.1 | XP_008769489.1 | 5 - 6 | 984 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |