Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770708.1 | XP_008768930.1 | 11 - 12 | 2134 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |