Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039336.1 | NP_001034425.1 | 7 - 8 | 771 | 111 | |
GenBank mRNA | AY886539.1 | - | 7 - 8 | 771 | 111 | |
BC098018.1 | - | 7 - 8 | 735 | 111 | ||
BC127440.1 | - | 7 - 8 | 781 | 7456492 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |