Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017240.2 | NP_058936.1 | 6 - 7 | 742 | 87 | |
XM_006251951.2 | XP_006252013.1 | 8 - 9 | 892 | 87 | ||
GenBank mRNA | X15939.1 | - | 6 - 7 | 744 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |