Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pspc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025672.1 | NP_001020843.1 | 1 - 2 | 514 | 98 | |
XM_006252065.2 | XP_006252127.1 | 1 - 2 | 556 | 98 | ||
GenBank mRNA | BC099204.1 | - | 1 - 2 | 514 | 7384173 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |