Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017257.2 | NP_058953.1 | 3 - 4 | 146 | 90 | |
GenBank mRNA | BC058486.1 | - | 3 - 4 | 184 | 6918985 | 90 |
BC097331.1 | - | 3 - 4 | 146 | 7389180 | 90 | |
D45250.1 | - | 3 - 4 | 196 | 90 | ||
FQ226805.1 | - | 3 - 4 | 177 | 90 | ||
FQ227049.1 | - | 3 - 4 | 190 | 90 | ||
FQ232840.1 | - | 3 - 4 | 177 | 90 | ||
FQ233763.1 | - | 3 - 4 | 172 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |