Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153315.2 | NP_695227.2 | 3 - 4 | 539 | 105 | |
GenBank mRNA | AB062758.1 | - | 3 - 4 | 355 | 105 | |
BC070961.1 | - | 3 - 4 | 539 | 7099044 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |