Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168583.1 | NP_001162055.1 | 6 - 7 | 842 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |