Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030850.1 | NP_110477.1 | 5 - 6 | 701 | 60 | |
GenBank mRNA | AF038870.1 | - | 5 - 6 | 701 | 60 | |
BC078811.1 | - | 5 - 6 | 688 | 7130736 | 60 | |
U96133.1 | - | 5 - 6 | 690 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |