Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168000.1 | NP_001161472.1 | 8 - 9 | 889 | 84 | |
XM_006231924.2 | XP_006231986.1 | 10 - 11 | 984 | 84 | ||
XM_006231925.2 | XP_006231987.2 | 10 - 11 | 1391 | 84 | ||
GenBank mRNA | JQ013732.1 | - | 8 - 9 | 1835 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |