Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106400.1 | NP_001099870.1 | 14 - 15 | 1937 | 125 | |
XM_008760599.1 | XP_008758821.1 | 12 - 13 | 1579 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |