Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031707.1 | NP_113895.1 | 5 - 6 | 824 | 59 | |
XM_008760635.1 | XP_008758857.1 | 5 - 6 | 1392 | 59 | ||
GenBank mRNA | AB007688.1 | - | 5 - 6 | 831 | 59 | |
AB017140.1 | - | 5 - 6 | 824 | 59 | ||
AF093268.1 | - | 5 - 6 | 1261 | 59 | ||
AJ276328.1 | - | 5 - 6 | 544 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |