Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008372.1 | NP_001008373.1 | 11 - 12 | 1406 | 118 | |
XM_006231776.2 | XP_006231838.1 | 11 - 12 | 1396 | 118 | ||
XM_006231778.2 | XP_006231840.1 | 11 - 12 | 1433 | 118 | ||
XM_008760639.1 | XP_008758861.1 | 12 - 13 | 1509 | 118 | ||
GenBank mRNA | BC085771.1 | - | 11 - 12 | 1406 | 7191068 | 118 |
FQ211725.1 | - | 11 - 12 | 1425 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |