Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053375.1 | NP_445827.1 | 1 - 2 | 462 | 132 | |
GenBank mRNA | AF247450.1 | - | 1 - 2 | 462 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |