Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004208.1 | NP_001004208.1 | 3 - 4 | 488 | 123 | |
GenBank mRNA | BC079007.1 | - | 3 - 4 | 488 | 7113129 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |