Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769512.1 | XP_008767734.1 | 5 - 6 | 608 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |